Anti-IGF-IR (Ab-1) Mouse Monoclonal Antibody [clone: αIR3]

Supplier: MilliporeSigma
GR11-100UG GR11L-100UG
CA80000-474EA 1178.38 CAD
CA80000-474 CA80000-476
Anti-IGF-IR (Ab-1) Mouse Monoclonal Antibody [clone: αIR3]
Antibodies
Purified mouse monoclonal antibody generated by immunizing SJL mice with the specified immunogen and fusing splenocytes with FO mouse myeloma cells (see application references). Recognizes the ~130 kDa α and the ~90 kDa β subunits of IGF-I receptor.

Type: Primary
Antigen: IGF-IR (Ab-1)
Clonality: Monoclonal
Clone: αIR3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR