Anti-GFAP Mouse Monoclonal Antibody

Supplier: EMD Millipore
IF03L-100UG
CA80000-584EA 1158.42 CAD
CA80000-584
Anti-GFAP Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Glial Fibrillary Acidic Protein (GFAP) (G-A-5) Reacts with Chicken, Human, Pig (Porcine, Swine), Rat

Type: Primary
Antigen: Glial Fibrillary Acidic Protein (GFAP) (G-A-5)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Chicken, Human, Pig (Porcine, Swine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR