Anti-c-ErbB2/c-Neu (Ab-4) Mouse Monoclonal Antibody [clone: 7.16.4]

Supplier: MilliporeSigma
OP16-100UG OP16L-100UG
CA80001-274EA 1150.53 CAD
CA80001-274 CA80001-276
Anti-c-ErbB2/c-Neu (Ab-4) Mouse Monoclonal Antibody [clone: 7.16.4]
Antibodies
Recognizes the ~185 kDa c-ErbB2/c-Neu protein in overexpressing B104-1-1 cells.

Type: Primary
Antigen: c-ErbB2/c-Neu (Ab-4)
Clonality: Monoclonal
Clone: 7.16.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat, Human
Order Now

SPECIFICATIONS

Antigen symbol c-ErbB2/c-Neu (Ab-4) c-ErbB2/c-Neu (7.16.4)
Environmentally Preferable
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Clone 7.16.4
Reactivity Rat,Human Human,Rat
Host Mouse Mouse
Isotype IgG2a
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Storage temperature 2...8 °C
Shipping temperature Wet ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR