Anti-c-Fos (2G9C3) Mouse Monoclonal Antibody

Supplier: EMD Millipore
OP17L-200UG
CA80109-980EA 1159.57 CAD
CA80109-980
Anti-c-Fos (2G9C3) Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-c-Fos (Ab-1) (2G9C3) Reacts with Human, Mouse, Rat

Type: Primary
Antigen: c-Fos (2G9C3)
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR