Anti-CDH1 Mouse Monoclonal Antibody

Supplier: EMD Millipore
CC43-100UG
CA80511-828EA 1186.99 CAD
CA80511-828
Anti-CDH1 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Cdh1 (Ab-2) (DH01) Reacts with Human

Type: Primary
Antigen: Cdh1 (DH01)
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR