Anti-CTSB Mouse Monoclonal Antibody

Supplier: EMD Millipore
IM27L-100UG
CA80000-642EA 1232.39 CAD
CA80000-642
Anti-CTSB Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Cathepsin B (Ab-1) (CA10) Reacts with Cow (Bovine, Cattle), Human, Pig (Porcine, Swine)

Type: Primary
Antigen: CTSB
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR