Anti-TIMP3 Mouse Monoclonal Antibody [clone: 136-13H4]

Supplier: EMD Millipore
IM43-100UG
CA60801-074EA 1101.97 CAD
CA60801-074
Anti-TIMP3 Mouse Monoclonal Antibody [clone: 136-13H4]
Antibodies
Primary Mouse Anti-TIMP-3 (136-13H4) Reacts with Baboon, Human, Rabbit (Lapine)

Type: Primary
Antigen: TIMP3
Clonality: Monoclonal
Clone: 136-13H4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR