Anti-SYN1 Rabbit Polyclonal Antibody

Supplier: EMD Millipore
574777-10UG
CA80057-552EA 1197.32 CAD
CA80057-552
Anti-SYN1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Synapsin I Reacts with Cow (Bovine, Cattle), Human, Mouse, Rat

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR