Anti-RENT1 Goat Polyclonal Antibody

Supplier: EMD Millipore
DR1015-100UG
CA80603-982EA 1371.69 CAD
CA80603-982
Anti-RENT1 Goat Polyclonal Antibody
Antibodies
Primary Goat Anti-RENT1 Reacts with Human

Type: Primary
Antigen: RENT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR