Anti-Nitro-Tyr Mouse Monoclonal Antibody

Supplier: EMD Millipore
487923-50UG
CA80053-442EA 623.58 CAD
CA80053-442
Anti-Nitro-Tyr Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Nitrotyrosine (CC22.8C7.3) Reacts with Pan

Type: Primary
Antigen: Nitrotyrosine (CC22.8C7.3)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR