Anti-IP 3 Receptor (IPR.1) Mouse Monoclonal Antibody

Supplier: EMD Millipore
407140-50UG
CA80052-540EA 1342.54 CAD
CA80052-540
Anti-IP 3 Receptor (IPR.1) Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-IP 3 Receptor (IPR.1) Reacts with Cow (Bovine, Cattle), Dog (Canine), Human, Mouse, Pig (Porcine, Swine), Rabbit (Lapine), Rat

Type: Primary
Antigen: IP 3 Receptor (IPR.1)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Cow (Bovine), Dog (Canine), Human, Mouse, Pig (Porcine, Swine), Rabbit (Lapine), Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR