Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: EMD Millipore
374090-100UL
CA80602-922EA 1372.95 CAD
CA80602-922
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Heme Oxygenase-1 (1-30) Reacts with Dog (Canine), Hamster, Human, Monkey, Mouse, Rat

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 1-30 aa
Host: Rabbit
Isotype:
Reactivity: Dog, Hamster, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR