Anti-PTK2 Rabbit Polyclonal Antibody

Supplier: EMD Millipore
PS1008-100UL
CA80016-822EA 1527.97 CAD
CA80016-822
Anti-PTK2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-FAK (pTyr 861 ) Reacts with Chicken, Human, Mouse

Type: Primary
Antigen: FAK (pTyr 861 )
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR