Anti-CD28 Mouse Monoclonal Antibody [clone: ANC28.1/5D10]

Supplier: EMD Millipore
217669-100UG
CA80108-540EA 1065.04 CAD
CA80108-540
Anti-CD28 Mouse Monoclonal Antibody [clone: ANC28.1/5D10]
Antibodies
Primary Mouse Anti-CD28 (ANC28.1/5D10) Reacts with Human

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: ANC28.1/5D10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR