Anti-Catalase Rabbit Polyclonal Antibody

Supplier: EMD Millipore
219010-1ML
CA80050-886EA 451.73 CAD
CA80050-886
Anti-Catalase Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Catalase Reacts with Human, Mouse

Type: Primary
Antigen: Catalase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR