Anti-FLAG™ conjugated Proteins Mouse Polyclonal Antibody [clone: 29E4.G7]

Supplier: Rockland Immunochemicals
200-301-B13 200-301-B13 200-301-383 200-301-383
CA200-301-383EA 504 CAD
CA200-301-383 RL200-301-B13 RL200-301-383 CA200-301-B13
Anti-FLAG™ conjugated Proteins Mouse Polyclonal Antibody [clone: 29E4.G7]
Antibodies
Primary Mouse Anti-FLAG™ conjugated proteins Reacts with Pan

Type: Primary
Antigen: FLAG™ conjugated Proteins
Clonality: Polyclonal
Clone: 29E4.G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR