Anti-GPANK1 Mouse Monoclonal Antibody

Supplier: EMD Millipore
NB20-100UG
CA80000-938EA 1100.08 CAD
CA80000-938
Anti-GPANK1 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Ankyrin G (463) Reacts with Dog (Canine), Human, Mouse, Rat

Type: Primary
Antigen: Ankyrin G (463)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Dog (Canine), Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR