Nalgene® Polycarbonate Multipurpose Jars with Covers, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS5300-0507 DS5300-9609 DS5300-9910
36324-989CS 216.14 CAD
36324-989 36325-008 36325-356 36325-360
Nalgene® Polycarbonate Multipurpose Jars with Covers, Thermo Scientific
Jars General Use Jars
Nalgene® Polycarbonate Multipurpose Jars are useful as water baths, aquaria, terraria or freeze-drying chambers
Cat. Nos. DS5300-0504; DS5300-0507 may be used for vacuum service if not autoclaved. For vacuum service, see Cat. Nos. 5305 and DS5320.
You can drill, tap or apply clamps to attach heating or cooling equipment. The covers permit the jars to be stacked.