Finnpipette® Novus GLP Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
4700470 4700490 4701120 4701130
89201-930EA 2942.79 CAD
89201-930 89201-932 89201-934 89201-936
Finnpipette® Novus GLP Kits, Thermo Scientific
Pipettes Micropipettes
Finnpipette® Novus GLP Kits provide ergonomic and easy-to-use Finnpipette Novus pipetters in an all-inclusive kit containing everything you need for pipetting.
The Finnpipette® Novus Electronic Single Channel Pipettes offer performance with simple operation and reduced stress for comfortable, productive pipetting.
Pipettors feature 10 pipetting functions and nine pipetting speeds. Unique to Novus, the adjustable index-finger trigger action prevents common repetitive stress injuries caused by thumb-driven pipettes, and the easy-to-follow prompts guide Novus users through 10 pipettingMore Product Information