Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS0320-2545 DS0320-5033 DS0320-5045
28199-322EA 397.82 CAD
28199-322 28199-324 28199-325
Nalgene® Polysulfone Reusable Bottle Top Filters, Thermo Scientific
Filters Bottle Top Filters
Nalgene® Polysulfone Reusable Bottle Top Filters offer the convenience of a bottle top filter with the cost savings and waste reduction of a reusable filter housing.
The filters feature a removable sterilization membrane support plate designed to provide maximum flow rate and throughput, as well as a vacuum port on the lower half of the assembly and molded-in graduations. The polypropylene tubing adapter can be stuffed with cotton for sterile venting during autoclaving.
The deep threads screw securely onto the bottle for convenient filling and vacuum filtration.