Proteinase K
Supplier: VWR
E195-25ML
E195-5ML
97062-668EA
1644.87
CAD
97062-668
97062-670
Proteinase K
Enzymes
Proteinase K is a non-specific serine protease with a molecular weight of approximately 18 kDa.
This enzyme exhibits high stability and activity in the presence of SDS, EDTA, and urea, as well as over a wide pH range. Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range.