VWR® Phosphate Buffered Saline (PBS) 20X, Ultra Pure Grade
Supplier: VWR
E703-1L
E703-500ML
97062-948EA
170.8
CAD
97062-948
97062-950
VWR® Phosphate Buffered Saline (PBS) 20X, Ultra Pure Grade
Buffers
Cell Culture Buffers
1X Solution Composition:135 mM NaCl, 2.7 mM KCl,11 mM Phosphate Buffer.Does not contain magnesium or calcium.
- For making 1X solution dilute 1:20 in deionized water
- pH at 25°C (1X PBS): 7.4 – 7.6
- May be sterilized by filtration or autoclave, if desired