Fmoc-D-Pro-OH

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: N-Fmoc-D-proline

02064-25G 02064-5G
CH02064-25GEA 202.56 CAD
CH02064-25G CH02064-5G
Fmoc-D-Pro-OH
Fmoc-D-Pro-OH
Formula: C₂₀H₁₉NO₄
MW: 337.38
MDL Number: MFCD00077067
CAS Number: 101555-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR