HyLite® 47-402 Nitrile Gloves, Fully Coated, Ansell
Supplier: Ansell Healthcare
205940 205941
80092-598CS 2259.84 CAD
80092-598 80092-600 80092-604
HyLite® 47-402 Nitrile Gloves, Fully Coated, Ansell
Gloves
A versatile alternative to cotton, light leather, and PVC, this special nitrile formulation provides an excellent combination of flexibility, mechanical resistance and oil repellency.
  • Silicone-free
  • Comfortable hand shape
The natural hand shape is easier to wear for long periods.
Use in general industry, maintenance, janitorial services, sanitation, metal fabrication, oil processing and refining, petrochemical, public utilities, and logistics and warehousing industries. Uses include assembling and inspecting engine parts and small components;More Product Information