Indirect Vent Goggles, 551 Softsides

Supplier: Bouton
112-5190400B 112-5190400B
77531-604CS 1967.75 CAD
300076-136 77531-604
Indirect Vent Goggles, 551 Softsides
Goggles
Lenses made of polycarbonate block 100% of harmful UV Radiation.

  • Made in the U.S.A
  • Chemical and anti splash protection
  • Ventilation via hooded vents to keep liquids out
  • Elastic headband

Weight: 0.23 lbs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR