Puritan® Rovers® Cytology Collection Devices, Puritan Medical Products
Supplier: Puritan Medical Products
2196 2195COMBI 2196
CA10808-150CS 4189.13 CAD
CA10808-150 CA10808-166 CA95056-638 95056-636 95056-638 10808-150
Puritan® Rovers® Cytology Collection Devices, Puritan Medical Products
Swabs Swabs for Sampling
These brushes are used to collect cervical cells for conventional Pap smears or in conjunction with the monolayer prep process
The Cervex-Brush® Combi replaces the middle bristles of the Cervex-Brush® with a complete endocervical brush to allow for simultaneous Pap smear collection of ectocervical and endocervical cells with a single device insertion. These brushes should not be used during pregnancy.
The EndoCervex-Brush® provides 600 separate collection "bristles" for the simultaneous collection of cellular material. The Brush features a stiff yet flexible bristle head with a protective tip to enhance the overall collection process. A safety ring just below the bristle head will decrease the possibility of the brush entering the endometrium during collection. The brush head is detachable after collection.
The Cervex-Brush® has semi-circular polyethylene bristles of various lengths to collect endocervical, ectocervical, and transformation zone cells without the use of a spatula.
Certifications: CE marked for medical use. Made in the USA.