Explorer® Analytical Balances, Ohaus®
Supplier: Ohaus
83021331 83021332 83021335 83021334 83021337
89171-446EA 9684.52 CAD
89171-446 89171-448 89171-450 89425-976 89425-978
Explorer® Analytical Balances, Ohaus®
Balances and Scales
A large color touch-screen display, four programmable touch-free sensors, and icon-driven app software make the Explorer the easiest to use, most advanced balance in its class today.
  • Intuitive operation
  • Intelligent calibration
  • Ingenious modular design
All New Explorers come standard with AutoCal™, OHAUS’ automatic internal calibration adjustment system
The base and terminal separate for maximum flexibility in all work areas. The design also isolates the weighing chamber to enhance stability and speeding up results.
Results are always fast and accurate.
Certifications: Agency Approvals CE, FCC, CMC, OIML Class I and Class II.
Ordering information: Accessories not included. Draftshield is a standard Accessory. Tower Mount is an optional Accessory. Switching AC adapter with multiple plug head type for country, Weighing pan, Pan support, Multilanguage operator manual with quick start guide and In use Cover - Terminal are included in box. Two year warranty. Model EX224N is NTEP certified legal for trade.