jetPEI®, HTS DNA Transfection Reagent
Supplier: SARTORIUS STEDIM NORTH AMERICA INC.
101000053 101000020 101000053
CA89129-916EA 824.35 CAD
CA89129-916 CA89129-918 89129-916
jetPEI®, HTS DNA Transfection Reagent
Transfection Reagents
jetPEI® transfection reagent is a linear polyethylenimine derivative, free of components of animal origin, providing highly effective and reproducible gene delivery, perfectly suited for High Throughput Screening (HTS) applications.
  • Well-suited for adherent and suspension cells
  • Fast and efficient methods to transfect cells for HTS
  • Compatible with serum and antibiotics
  • Exceptionally reproducible results
  • Reverse, batch & forward protocols available
jetPEI® transfection reagent is particularly well suited for automated or manual High Throughput Screening (HTS) with three protocols available: Reverse, batch and forward.
Certifications: Since 2002, the company is ISO 9001 certified and since then this certification has been successfully renewed every three years.
Every batch of jetPEI® is tested in-house by DNA transfection of HeLa cells. Transfection with a firefly Luciferase gene under the control of CMV promoter gives at least 109 RLU (relative light unit)/mg of protein. The value for each batch is indicated on the Certificate of Analysis.
Delivery information: 150 mM NaCl solution is included. 1 mL of jetPEI® transfection reagent is sufficient to perform 2000 transfections in 96-well plates