Nalgene® Polypropylene Economy Bottles, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
2087-0001 2087-0002 2087-0004 2087-0008
89096-750CS 156.26 CAD
89096-750 89096-752 89096-754 89096-756 89096-758
Nalgene® Polypropylene Economy Bottles, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
  • Bottles are the economical, quality choice for lighter-duty lab use
  • Priced below low-quality plastic or glass bottle brands
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent polypropylene copolymer. Like most Nalgene bottles, the Economy Bottles are autoclavable, guaranteed leakproof and are made in our ISO-certified USA manufacturing facility.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Thermo Scientific Nalgene High-Density Polyethylene Environmental Sample Bottles are also available; contact your Avantor sales representative for more information.