KleenGuard™ V90 Shield Safety Goggles with Face Shield, Kimberly-Clark Professional
Supplier: Ansell Healthcare
18629 18629
89130-975CS 435.36 CAD
89130-975 CA89130-975
KleenGuard™ V90 Shield Safety Goggles with Face Shield, Kimberly-Clark Professional
Goggles
The safety goggles feature KleenGuard™ Monogoggle™ XTR goggles integrated with a face shield to provide full face protection from liquids and particulates without the hassle of primary eye protection underneath a face shield.
  • Indirect ventilation provides increased airflow, minimizing condensation and fogging
  • Curved face shield conforms to the shape of the user's face, and is a splash and impact protector
  • Soft, pliable material around the eyes and nose ensure a comfortable fit
  • Goggles can be used over most prescription eyeglasses
  • Shield is detachable
More Product Information