Corning® Gosselin™ Dippers with Removable Handles, Corning
Supplier: Corning
TAFL1-01 TAT52C-01
89200-976CS 584.03 CAD
89200-976 10852-182
Corning® Gosselin™ Dippers with Removable Handles, Corning
Samplers Sample Dippers
Corning® Gosselin™ offers a wide range of disposable sampling tools including Dippers with Removable Handles
  • Shock resistant
  • Assured sterile 10-3
Corning® Gosselin™ dipper consists of a blue polypropylene container with a polyethylene removable handle. The 90 mL sized container has a hinged cap; all other sizes have screw caps.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food & Beverage, Environmental/Water Testing, etc.
See also Corning® Gosselin™ Cutlery, BlenderMore Product Information