Corning® Gosselin™ Blender Bags, Corning
Supplier: Corning
FBAG-03 FBAG-04 BBAG-03 BBAG-04 SM1-01 SM2-01 SM3-01 CLIP-01
89200-858CS 1670.53 CAD
89200-858 89200-860 89200-862 89200-864 89200-866 89200-868 89200-870 89200-872
Corning® Gosselin™ Blender Bags, Corning
Bags Blender Bags
These polyethylene sterile blender bags have a lateral or full filter and are commonly used for dairy industry products.
  • Assured sterility 10⁻³
  • Lot number provided on each filter bag
  • Non-woven filter provides for stronger filtration power
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing and more.
See also Corning® Gosselin™ dippers with removable handles, cutlery, pipets, and inoculating loops and spreaders.
Certifications: Food contact : Raw material is compliant with regulation EC N°1935/2004
Sterility Assurance Level based on ISO 11137 : STERILE SAL 10⁻³
Ordering information: The blue and white clips for the blender bags are sold separately.