Straight Blade Impellers, Caframo
Supplier: Caframo
A131 A231 A131 A163 A163
CA58960-142EA 174.87 CAD
CA58960-142 CA58960-200 52428-770 52428-776 CA52428-776
Straight Blade Impellers, Caframo
Stirrers Stirring Tools
The model A131 radial flow impeller is effective for high speed, low viscosity mixing
The model A163 straight blade of 40 mm (11/2") diameter, is composed of electropolished 316L stainless steel. The 8 mm (5/16") bore is compatible with all Caframo shafts. Set screws included. For high shear, low viscosity mixing with moderate vortexing, this blade will vortex 19 L (5 gal.) of water at 1200 rpm.
The model A231 radial flow impeller is effective for high speed, low viscosity mixing. The 30 mm (11/4”) diameter blade is welded to an 8 mm (5/16") diameter shaft, all electropolished 316L stainless steel. A high shear impeller with moderate vortexing properties.
The 50 mm (2") diameter blade is welded to an 8 mm (5/16") diameter shaft, all electropolished 316L stainless steel. A high shear impeller with moderate vortexing properties, creating a vortex in 19 L (5 gal.) of water at 1200 rpm.