High Torque Overhead Stirrers, Caframo
Supplier: Caframo
BDC3030 BDC6015 BDC6015 BDC1850
CA58960-165EA 2886.61 CAD
CA58960-165 77776-354 CA58960-170 CA58960-180
High Torque Overhead Stirrers, Caframo
Stirrers Overhead Stirrers
Designed for continuous, high viscosity mixing
These benchtop overhead stirrers are most suitable for highly viscous mixing applications. Other applications include cosmetics and personal care products, inks and coatings, paint, pharmaceuticals, pulp and paper, pilot plants, adhesives, and epoxies.
Brushless, sparkless DC motor and two speed transmission deliver constant performance under changing conditions. Speed is maintained through electronic control as product viscosity changes. Stirrers are built with a totally enclosed metal housing, through shaft, digital display, and chuckMore Product Information