FOCUS™ Extraction Buffers, G-Biosciences
Supplier: G-Biosciences
One of the most important considerations before running 2D gel electrophoresis is the choice of protein solubilization buffers
One way to minimize the risk of carbamylation is to prepare the urea based reagents fresh before each use. G-Biosciences has developed a series of dry urea based pre-mixed and ready-to-use solubilization buffers. Simply add an appropriate volume of the supplied rehydration buffer to the dry buffer and then use to solubilize proteins, saving time and improving the quality of IEF/2D gel electrophoresis. FOCUS™ Extraction Buffers are also designed to be used as rehydration buffers for IPG strips.
FOCUS™ Extraction Buffers are experimentally optimized concentrations of critical agents, buffering and stabilizing agents, including urea, thiourea, Nonidet® P-40, CHAPS, and sulfobetaines (SB). The FOCUS™ Extraction Buffers are designed to produce optimal protein extraction and improved spot resolution for 2D gel analysis.
A range of FOCUS™ Extraction Buffers have been developed (see table) and depending on the nature of the samples, one or more of the buffers suitable for your applications can be ordered. FOCUS™ Extraction Buffer-I is suitable for most applications, however for stronger solubilization effects, we recommend FOCUS™ Extraction Buffer-II, III, IV, V or VI. For analysis of a proteome, a single buffer may not be suitable and sequential solubilization using different FOCUS™ Extraction Buffers will help in identifying new proteins. For analysis of a proteome, a single buffer may not be suitable and sequential solubilization using different FOCUS™ Extraction Buffers will help in identifying new proteins.
The suitable buffer must solubilize proteins effectively, without disturbing the native charge of the proteins. Urea, a common chaotrope, is widely used for solubilization and denaturation of proteins. One of the disadvantages of using urea is carbamylation. Urea in water exists in equilibrium with ammonium cyanate, the level of which increases with increasing temperature and pH. Cyanate reacts with α-amino and ε-amino groups of proteins and induces a change in the isoelectric point of proteins. This leads to artifactual results and therefore carbamylation must be avoided.
Ordering information: Accessories are available.