HistoGel™ Specimen Processing Gel, Epredia
Supplier: Epredia
HistoGel™
HGSK-2050-1 HG-4000-144 HG-4000-012 HG-4000-012 HGSK-2050-1 HG-4000-144 WB-4010-1 WB-4010-1
CA89126-234EA 7548.31 CAD
CA89126-234 83009-992 CA16002-560 CA83009-992 83009-994 CA83009-994 89126-234 16002-560
HistoGel™ Specimen Processing Gel, Epredia
Stains Microscopy and staining reagents
Encapsulate and retain even small, friable and viscous specimens during histological processing with Epredia™ HistoGel™ Specimen Processing Gel.
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
This gel encapsulates and retains specimens during histological processing, regardless of the specimens’ small, friable, or viscous nature. Chemical agents penetrate the specimen without allowing it to escape from the gel. The gel does not retain histological stains, eliminating unwanted discoloration around specimens on slides. It is virtually unnoticeable during sectioning and will not pop out of the paraffin block during cutting.
Ordering information: Starter Kit includes twelve 10ml (0.34oz.) HistoGel™ vials, two WonderBlock cooling blocks, a dry bath incubator, and 500 disposable pipets.