Signature Series™ Bluing Reagent, Epredia
Supplier: RICHARD-ALLAN SCIENTIFIC CA
Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series™ Bluing Reagent.
- Specially formulated
- Prevents tissue separation from the slide
- One-minute water rinse
- Five liter packaging saves 35% more storage space
This specially formulated bluing reagent prevents tissue separation from the slide. It requires only a one-minute water rinse versus conventional bluing reagents which require up to five minutes. Buffered to ensure proper alkalinity (pH = 8), reduce messy stain spills, and increase stain stability. The unique packaging saves 35% more storage space than traditional packaging while also reducing plastic by 9%.