Canisters, 95 kPa, Therapak®
Supplier: Therapak, LLC
23027 23072 23080
89170-956CS 634.94 CAD
89170-956 89170-958 89170-960
Canisters, 95 kPa, Therapak®
Mailers and Shippers
Designed for the international transport of specimens, these canisters can be used with primary containers, absorbent materials, and a rigid outer container such as a box to conform to IATA Packing Instruction 650 for Biological Substances.
The 5 tube canister is made of translucent PET material with a screw-cap closure and a special liner to ensure watertight transportation. The 12 tube container features an O-ring gasket that creates a tight seal for watertight transportation.
Order Now
SPECIFICATIONS
Environmentally Preferable