Prostate Biopsy Kits, Azer Scientific
Supplier: Azer Scientific
10-831 10-820 10-816 10-816
CA89129-906CS 2411.06 CAD
89129-904 89129-902 CA89129-906 89129-906
Prostate Biopsy Kits, Azer Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
These ready-to-use kits are packaged with 8, 12, or 16 vials, which are prefilled with 20 ml of 10% neutral buffered formalin and a leak-resistant seal.
  • Cardboard insert labeled with the appropriate section of the prostate
Each container is half-filled with 10% neutral buffered formalin.
The insert of each kit is labeled with the appropriate section of the prostate, which allows for easy tissue sampling.
Certifications: Vials are 95 kPA compliant.
Ordering information: Kits are also available with components, including a biohazard bag, formalin absorbent pad, and desiccant pack.
Packaging: Eight 20 ml Formalock™ containers are included. Vials are prefilled with 20 ml of 10% neutral buffered formalin and a leak-resistant seal. Each case contains 25 kits.