Corning® Gosselin™ Straight Containers, Corning
Supplier: Corning
FL300-04 FL60-04 FL100-05
89200-908CS 450.34 CAD
89200-908 89200-910 89200-912 89200-918
Corning® Gosselin™ Straight Containers, Corning
Sample Containers General Purpose Containers
Made of PP, natural or colored.
  • Available in sterile or under aseptic conditions packed versions
  • Very good chemical and temperature resistance
  • Graduated
  • Without cap
Ideal for sampling, storage and transport of liquid and solid samples.
Sized to provide adequate volume for milk sampling. It is shock resistant and is assured sterile 10-3.
Corning® Gosselin™ is a Corning brand of laboratory plasticware designed for use in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.