Superfrost® Plus and Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Supplier: Erie Scientific
9951BOPLUS-006 9951LPLUS-006 9951TPLUS-006 9951GLPLUS-006 9951APLUS-006 4951PLUS-4-006
89082-258CS 2172.03 CAD
89082-258 89082-260 89082-262 89082-264 89082-266 95057-985
Superfrost® Plus and Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Slides Adhesive Slides
Slides are treated to electrostatically attract frozen tissue sections and cytology preparations, eliminating the need for adhesives or coating steps.
Order Now
SPECIFICATIONS
Environmentally Preferable