Pt 100 Precision Digital Thermometers, Thermco
Supplier: Thermco
ACCD370PFC ACCD370PUFC ACCD370P ACCD370PDFC ACCD370PWFC
89137-110EA 1454.36 CAD
89137-110 89137-112 10145-558 10145-560 10145-562
Pt 100 Precision Digital Thermometers, Thermco
Thermometers Digital Thermometers
Precision RTD digital thermometers can be used as a temperature standard for the verification and calibration of laboratory equipment, as well as liquid-in-glass, digital, and dial bi-metal thermometers
Records minimum, maximum, and average temperatures. Devices feature a 7-segement LCD display and backlight. Waterproof
Certifications: Certified and traceable to NIST standards. Pt100 Platinum according to EN 60751.
Ordering information: Includes insertion probe and three AAA batteries. Other certified temperatures can be requested; please contact your Avantor representative for more information.