Triple Display Certified Thermometers, Thermco
Supplier: Thermco
ACC895FRE ACC895REF ACC895INC ACC895BLK ACC895AMB ACC895WB
89137-068EA 194.29 CAD
89137-068 89137-070 89137-072 89137-074 10025-456 10025-464
Triple Display Certified Thermometers, Thermco
Thermometers Digital Thermometers
These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously
The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument. Thermometers also feature a flip-open stand, wall mounting key hole, and an AAA alkaline battery.
An audible alarm sounds when the temperature rises above or below a set temperature. The minimum/maximum feature monitors critical temperature for any period of time
Certifications: Each thermometer is certified and traceable to NIST. Each thermometer is serialized.