Shandon™ Neoprene Cork Dissecting Boards, Thermo Scientific
Supplier: Epredia
36119 361
41799-048EA 427.48 CAD
41799-048 41799-046
Shandon™ Neoprene Cork Dissecting Boards, Thermo Scientific
Dissecting Boards
Work on a durable, cut-resistant surface over a marine plywood base with Shandon™ Neoprene Cork Dissecting Boards.
  • Blended rubber and cork composition resists absorption and allows for thorough rinsing
  • Rubber bumpers attached to each corner raise the cutting surface to 3.8 cm (1.5")