Puritan HydraFlock® Flocked Swabs, Puritan Medical Products
Supplier: Puritan Medical Products
3406-H 25-3406-H 25-3306-H 25-3316-H 25-3317-H 25-3318-H 25-3506-H 25-3706-H 25-3606-H 25-3806-H 25-3206-H
89194-860CS 1103.29 CAD
89194-860 89194-862 89194-870 89194-872 89194-874 89194-876 89194-878 89428-646 89428-650 89497-312 89497-314 10124-664 10124-698 75788-568
Puritan HydraFlock® Flocked Swabs, Puritan Medical Products
Swabs Swabs for Sampling
Specifically engineerd for a wide range of bioloigcal specimens, HydraFlock®'s unique three-dimensional design facilitates greater specimen collection and complete elution
HdyraFlock® swabs are ideal for a variety of clinical settings and diagnostic tests.
Made in the USA.
These multi-length, split fibers have a greater absorption surface area than traditional perpendicular nylon flock fibers, thus allowing more rapid elution and sample preservation.
Certifications: CE marking for medical use. Patented: United States, Canada, Europe, Australia, Japan, and China. Patents Pending: Hong Kong.
Ordering information: Sterile swabs are individually wrapped. Nonsterile swabs are bulk packaged.