TRACK-IT™ Temperature Logger, Monarch Instrument
Supplier: Monarch Instrument
5396-0101 5396-0102 5396-0103 5396-0104 5396-0101-CAL 5396-0102-CAL 5396-0103-CAL 5396-0104-CAL
89184-528EA 219.07 CAD
89184-528 89184-530 89184-532 89184-534 89409-774 89409-776 89409-778 89409-780
TRACK-IT™ Temperature Logger, Monarch Instrument
Data Loggers
This Logger is a battery powered stand alone device that records up to 64000 samples of temperature data.
  • Push button or program to start recording
  • Four digit LCD display
  • Two user progammable alarms, high or low
  • Indication of alarms, triggered, small compact size
  • N.I.S.T. calibration available and MKT calculation included
  • High accuracy and repeatability
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery. The Track‐It™ Temperature Logger can be ordered with or without an LCD display and with a standard (one year) or long life battery (three year).
Information displayed on the LCD is also user programmable and can be sequenced by pressing the logger button. Both models have a multicolored LED indicator to notify user of alarms or general activity. A splash proof protective cover protects the logger when in use.
The unit is easily configured using the included Track‐It™ Software. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed.
Coin cell option: Battery: Lithium CR2032
Hi energy option: Battery: EF651625
Certifications: CE Marked.
Ordering information: Interactive Software and splash proof protective cover included.