SafetySpace™ Filter Pipette Tips, Sterile
Supplier: Sartorius
790011F 790021F 790101F 790201F 790301F 791001F 791211F 783201 790011F 790021F 790101F 790201F 790301F 791001F 791211F 783201 LH-795001F LH-XF780201 LH-XF781001 LH-XF791001
89092-962PK 386.76 CAD
89092-962 89092-964 89092-966 89092-968 89129-836 89129-838 89129-840 53495-410 10033-712 CA89092-962 CA89092-964 CA89092-966 CA89092-968 CA89129-836 CA89129-838 CA89129-840 CA53495-410 CA76416-014 CA76416-024 CA76416-026 CA77745-762
SafetySpace™ Filter Pipette Tips, Sterile
Pipette Tips
SafetySpace™ filter tips, made of virgin polypropylene, feature filter barriers that effectively capture solid and liquid aerosol particles.
  • Filter minimizes the risk of aerosol contamination
  • Large empty volume between the sample and filter prevents liquid from contacting the filter
  • Covers tip volumes from 10 to 1200 µl
  • Certified free of DNase, RNase, and endotoxins
  • E-beam pre-sterilized
  • Full traceability
  • Color-coded trays indicate the matching color-coded Sartorius pipette
The unique feature of SafetySpace™ filter tips is the additional space left between the sample and the filter that conventional filter tips do not provide. This extra space prevents the liquid from touching and permeating the filter, and thus guarantees pipetting accuracy. Any liquid types and pipetting techniques can be applied without the risk of liquid permeating the filter.
The filter is made of polyethylene without self-sealing additives to avoid any interference with the sample and results. The filter not only protects the sample against contamination, but also prolongs the maintenance interval of the pipette.
Packaging: Single tray rack. 96 tips in convenient and reusable tray racks (sales unit contains 10 tray racks, total 960 tips).