TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Supplier: Monarch Instrument
5396-0301 5396-0302 5396-0303 5396-0304 5396-0301-CAL 5396-0302-CAL 5396-0303-CAL 5396-0304-CAL 5396-0305 5396-0305-CAL
89184-584EA 1140.27 CAD
89184-584 89184-586 89184-588 89200-716 89409-792 89409-794 89409-796 89409-798 89409-800 89409-802
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Data Loggers
The pressure/temperature data logger is a battery powered stand alone water tight compact data logger that records up to 64000 samples of pressure and/or temperature data
The logger is packaged in a rugged submersible 316L stainless steel housing and has a standard ¹/₄" NPT fitting. The replaceable internal lithium battery has up to a 5 year life. The unit is easily configured using the included Track‐It™ software. Simply connect a standard USB cable to the logger and an open USB port on your PC and the Track‐It™ software automatically identifies the logger.
Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
It can be configured to record pressure and temperature or either parameter alone to maximize data storage space. The Track‐It™ pressure/temperature data logger can be ordered in four pressure ranges: 0 - 35, 0 - 150, 0 - 350 and 0 - 550 psi.
Certifications: CE marked.