TRACK-IT™ AC Event Logger, Monarch Instrument
Supplier: Monarch Instrument
5396-0711 5396-0712
89184-570EA 395.95 CAD
89184-570 89184-572
TRACK-IT™ AC Event Logger, Monarch Instrument
Data Loggers
The AC event data logger is a battery powered stand alone compact data logger that records up to 64000 on/off cycles.
Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours or can be set to store a sample at every event. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
The Track‐It™ AC event logger can also be used to record ambient temperature along with on/off cycles. This can help diagnose heat vs. cycle time issues. Information displayed on the LCD is also user programmable and can be sequenced by pressing the logger button. The multicolor LED indicator notifies the user of alarms, or general activity.
Use the Track‐It™ AC event logger to monitor cycle patterns of pumps, blowers and other electrical equipment powered by 120 or 240 Vac. The unit is easily configured using the included Track‐It™ software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger.
Ordering information: Interactive Software included.