TRACK-IT™ DC Voltage and DC Current Logger, Monarch Instrument
Supplier: Monarch Instrument
5396-0511 5396-0512 5396-0513 5396-0514 5396-0515 5396-0521 5396-0522 5396-0523 5396-0524 5396-0525 5396-0511-CAL 5396-0512-CAL 5396-0513-CAL 5396-0514-CAL 5396-0515-CAL 5396-0521-CAL 5396-0522-CAL 5396-0523-CAL 5396-0524-CAL 5396-0525-CAL
89184-548EA 359.61 CAD
89184-548 89184-550 89184-552 89184-554 89184-556 89184-558 89184-560 89184-562 89184-564 89184-566 89409-804 89409-806 89409-808 89409-810 89409-812 89409-814 89409-816 89409-818 89409-820 89409-822
TRACK-IT™ DC Voltage and DC Current Logger, Monarch Instrument
Data Loggers
The DC voltage and current data loggers are two channel battery powered stand alone compact data loggers that record up to 32000 samples each of DC voltage or current data
The unit is easily configured using the included Track‐It™ Software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. After the logger has been programmed, disconnect it from the PC and attach the voltage or current module.
The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
Units are available in 4 DC voltage ranges or for DC current. The loggers come with and LCD display with the option of a standard (1 year) or long life battery (3 year).
Certifications: CE marked.
Ordering information: Interactive software included.